Triacetoxyvinylsilane

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Vinylsilanetriyl triacetateTriacetoxyvinylsilaneVinyltriacetoxysilaneTriacetoxy(vinyl)silane

14-9250-50G 14-9250-250G
100204-914EA 46.71 USD
100204-914 100203-964
Triacetoxyvinylsilane
Triacetoxyvinylsilane
Formula: C₈H₁₂O₆Si
MW: 232.27 g/mol
Boiling Pt: 115 °C (10 mmHg)
Density: 1,167 (20°)
MDL Number: MFCD00014975
CAS Number: 4130-08-9
EINECS: 223-943-1
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR