Trimethylsilyl trifluoromethanesulfonate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Trimethylsilyl trifluoromethanesulfonate
, Trimethylsilyl trifluoromethanesulphonate
14-8600-25G
14-8600-5G
100200-410EA
178.34
USD
100200-410
100198-584
Trimethylsilyl trifluoromethanesulfonate ≥97%
Trimethylsilyl trifluoromethanesulphonate
Formula:
C₄H₉F₃O₃SSi MW: 222.26 g/mol Boiling Pt: 77 °C (80 mmHg) Density: 1,150 Flash Pt: 78 °F |
MDL Number:
MFCD00000406 CAS Number: 27607-77-8 EINECS: 248-565-4 UN: 2920 ADR: 8,II Merck Index: 12,09849 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...