Dimethoxy(diphenyl)silane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dimethoxydiphenylsilane
, Diphenyldimethoxysilane
14-2500-50G
14-2500-250G
100198-120EA
215.14
USD
100198-120
100196-044
Dimethoxy(diphenyl)silane ≥97%
Dimethoxy(diphenyl)silane
Formula:
C₁₄H₁₆O₂Si MW: 244.37 g/mol Boiling Pt: 161 °C (15 mmHg) Density: 1,0771 Flash Pt: 250 °F |
MDL Number:
MFCD00025690 CAS Number: 6843-66-9 EINECS: 229-929-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...