tert-Butylchlorodimethylsilane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
t-Butyldimethylchlorosilane
, tert-Butyldimethylsilyl chloride
, Chloro-tert-butyldimethylsilane
, tert-Butyldimethylsilyl Chloride
, TBDMS chloride
, tert-Butyldimethylchlorosilane
14-1775-10G
14-1775-50G
100209-652EA
77.85
USD
100209-652
100209-654
tert-Butylchlorodimethylsilane ≥97%
tert-Butylchlorodimethylsilane
Formula:
C₆H₁₅ClSi MW: 150.72 g/mol Boiling Pt: 125 °C Melting Pt: 86…89 °C Density: 0,81 g/cm³ (20 °C) Flash Pt: 73 °F |
MDL Number:
MFCD00000501 CAS Number: 18162-48-6 EINECS: 242-042-4 UN: 2925 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...