Sodium tetraethylborate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
11-0575-1G
11-0575-25G
11-0575-5G
100203-944EA
107.57
USD
100203-944
100203-946
100203-948
Sodium tetraethylborate ≥98%
Sodium tetraethylborate
Formula:
C₈H₂₀BNa MW: 150.05 g/mol Melting Pt: Dec. |
MDL Number:
MFCD00061547 CAS Number: 15523-24-7 UN: 3393 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...