Anti-IgG Goat Polyclonal Antibody (Cy5®)

Supplier: SeraCare
5230-0429
95058-948EA 294.98 USD
95058-948
Anti-IgG Goat Polyclonal Antibody (Cy5®)
Secondary Goat Anti-IgG (H+L) F(ab')2, Reacts with Human

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR