Anti-IgG gamma Goat Polyclonal Antibody (DyLight® 488)

Supplier: SeraCare
5230-0380
95058-490EA 469.77 USD
95058-490
Anti-IgG gamma Goat Polyclonal Antibody (DyLight® 488)
Secondary Goat Anti-IgG (gamma) Reacts with Human

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR