Micro 12™ Micro Centrifuge, Separation Technology
Supplier: Separation Technology
Micro 12™
100-152
89184-114EA 2015.05 USD
89184-114
Micro 12™ Micro Centrifuge, Separation Technology
Micro 12™ is a high perfomance microcentrifuge
Its versatile speed range is ideal for clinical, molecular biology, cell biology and bacteriology use. It is perfect for a variety of applications including quick spin downs, and cell and bacteria separations, microfiltrations, and small volume blood and urine separations. The rotor accomodates all tubes up to 2ml.