Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
Powder/Bead Dispensers
https://us.vwr.com/stibo/smallweb/std.lang.all/53/36/18945336.jpg
Adjustable powder dispensers utilize positive displacement for extremely precise placement delivery. Acting similar to a pipettor, depressing the plunger traps desired material in the tip. The material density will determine the mass transferred. The simple transporting operation minimizes powder waste and unnecessary replacement costs. Cost efficient manual models have an ergonomically designed handle and removable probes varying in millimeter diameter sizes. The easy-to-use powder dispensers ensure air pockets do not occur and may come calibrated.
SearchPresentationType-VERTICAL
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  HF Scientific
Description:   This portable dispenser accurately delivers the correct volume of DPD reagent for free or total chlorine