VWR® READY-PACK™ TG-SDS, 10X. Ultra-Pure Grade
Supplier: Avantor
0147-40L
97061-084CS 843.18 USD
97061-084
VWR® READY-PACK™ TG-SDS, 10X. Ultra-Pure Grade
Buffers Electrophoresis Buffers Running Buffers
TG-SDS buffer for preparation of Laemmli SDS-PAGE running buffer..
The 40 L pack size prepares 40 liters of 1X Tris-Glycine-SDS solution (0.025M tris base/0.192 M glycine/0.1% SDS). Each Ready-Pack prepares 1 liter of a 10X concentrate.