You Searched For: 4-Fluoro-7-nitrobenzofurazan


8  results were found

SearchResultCount:"8"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (TCA5593-100MG)
Supplier: TCI America
Description: [for HPLC Labeling] CAS Number: 29270-56-2 MDL Number: MFCD00010196 Molecular Formula: C6H2FN3O3 Molecular Weight: 183.10 Purity/Analysis Method:...

Supplier: Thermo Scientific Chemicals
Description: 4-FLUORO-7-NITROBENZOFURAZAN 98% 5MG

Catalog Number: (89139-596)
Supplier: Biotium
Description: is nonfluorescent until it reacts with primary or secondary amines to produce a fluorescent product. NBD fluoride has been extensively used as a derivatizing reagent for chromatography analysis of amino acids and low molecular weight amines. NBD fluoride is much more reactive than NBD chloride (90045) towards amine.


Catalog Number: (AAJ61336-MC)
Supplier: Thermo Scientific Chemicals
Description: Useful for amino acid and amine fluorescent labeling in HPLC

Supplier: Adipogen
Description: NBD fluoride is similar to NBD chloride except that the former is much more reactive toward amines. Both reagents are nonfluorescent until they react ...

Catalog Number: (101784-702)
Supplier: Matrix Scientific
Description: MF=C6H2Fn3O3 MW=183.10 Cas=29270-56-2 MDL=MFCD00010196 100Mg


Catalog Number: (76481-024)
Supplier: AAT BIOQUEST INC
Description: NBD-F has properties and applications similar to those of NBD-Cl.

Minority or Woman-Owned Business Enterprise Small Business Enterprise


Catalog Number: (77002-436)
Supplier: Ambeed
Description: 4-Fluoro-7-nitrobenzofurazan, Purity: 95%, CAS number: 29270-56-2, Appearance: Yellow to brown powder or crystals, Storage: Inert atmosphere, Store in freezer, under -20C, Size: 100MG

New Product


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8 of 8
no targeter for Bottom