    Sort by:
  • Search Within Results
(R)-(+)-4-Bromo-α-methylbenzylamine hydrochloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   (R)-(+)-1-(4-Bromophenyl)ethylamine hydrochloride, Purity: 97%, 99+% ee, CAS Number: 64265-77-6, Molecular weight: 236.54, Molecular formula: C8 H10 Br N. H Cl, Color: White to light yellow, Form: Powder, Size: 25g