Would you like to log in again?
Altair® 4X Multigas Detectors, MSA
Supplier: ORS NASCO, INC.
Altair®
454-10110443 454-10110444 454-10110445 454-10110446 454-10107602 454-10107603
  • Less calibration gas required with maximum 24 hours run time
  • Response and clear times less than 15 seconds
  • End-of-sensor-life warning allows for planned sensor replacement reducing down time
  • Galaxy™ System-compatible for powerful calibration and fleet-management system
  • MotionAlert™ feature activates when no movement is detected for 30 seconds, indicating "Man down"
  • InstantAlert™ feature is a manually activated alarm to alert others of hazardous situations
  • Bump check mode provides clear indication of bump check status and provides long-term record storage
  • Test to IP67
Certifications: UL, Class I, Div. 1, Groups A B, C, D, Class II, Div. 1 Groups F & G. T4, Tamb= -20 to 50 C, Tested to IP67.
ORDER
Expand All / Collapse All
Detectable Gas Supplier No. VWR Catalog Number Unit Price Quantity
LEL, Oxygen, Carbon Monoxide, Charcoal 454-10110443 300008-971 Each Retrieving
LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide, Charcoal 454-10107602 300008-969 Each Retrieving
LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide, Phosphorescent 454-10107603 300008-970 Each Retrieving
LEL, Oxygen, Carbon Monoxide, Phosphorescent 454-10110445 300008-973 Each Retrieving
LEL, Oxygen, Charcoal 454-10110444 300008-972 Each Retrieving
LEL, Oxygen, Phosphorescent 454-10110446 300008-974 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Sound Level 95 dB
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty