Ultra-Lok™ Self Retracting Lifelines, DBI-SALA, ORS Nasco
Supplier: ORS Nasco
Ultra-Lok™
098-3504450 098-3504451
300075-636EA 2218.03 USD
300075-636 300075-638
Ultra-Lok™ Self Retracting Lifelines, DBI-SALA, ORS Nasco
Fall Protection Fall Arresters
  • Limits arresting forces to 900 lbs or less
  • Reserve lifeline retention system for added safety
  • Self-locking swivel snap hook with impact indicator
  • Stainless steel working components for corrosion resistance
  • Swiveling anchorage loop to prevent twisting of lifeline