Ultra-Lok™ Self Retracting Lifelines, DBI-SALA, ORS Nasco
Supplier: ORS Nasco
Ultra-Lok™
098-3504431 098-3504433 098-3504430
300075-634EA 1237.83 USD
300075-634 300058-322 300058-324
Ultra-Lok™ Self Retracting Lifelines, DBI-SALA, ORS Nasco
Fall Protection Fall Arresters
  • Stainless steel working components for corrosion resistance
  • Limits arresting forces to 900 lbs or less
  • Reserve lifeline retention system for added safety
  • Self-locking swivel snap hook with impact indicator
  • Swiveling anchorage loop to prevent twisting of lifeline