Talon® Tie-Off Self Retracting Lifelines, DBI-SALA, ORS Nasco
Supplier: ORS Nasco
098-3102000
300058-310EA 603 USD
300058-310
Talon® Tie-Off Self Retracting Lifelines, DBI-SALA, ORS Nasco
  • 100% 2-leg tie-off protection and limit arresting forces to 900 lb
  • Also available with small or large steel hooks
  • Impact indicator on web and i-Safe™ technology for easy tracking of inspections and inventory
  • Includes aluminum rebar style hooks (2-1/4" opening) with ANSI compliant 3,600 lb gate
  • This lightwieght, compact SRL mounts directly to the back of most any full body harness