Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Talon® Self Retracting Lifelines, DBI/Sala

Supplier: ORS NASCO, INC.
Talon®
098-3101001 098-3101000
  • Also available with large gate hooks
  • Soft cover shock absorber for comfort
  • Includes easy to use swiveling self locking snap hook with new ANSI compliant 3,600 lb gate
  • Mounts directly to the back of most any full body harness
  • Web line to arrest falls within 42" while limiting arresting forces to 900 lb



 

ORDER

Expand All / Collapse All
Connection Type Hook Opening Material Load Capacity Length Weight Supplier No. VWR Catalog Number Unit Price Quantity
Self-Locking Swivel Snap, Ø 1" 3/4" Nylon Web 3600 lb [Max] 8 ft 3 lbs. 098-3101001 300058-308 Each Retrieving
Swivel Snaps, Black, Ø 1" with 1 Leg 3/4" Nylon Web 3600 lb [Max] 8 ft - 098-3101000 300075-622 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty