EZ Stop® III Shock Absorbing Lanyards, ORS Nasco
Supplier: ORS Nasco
EZ Stop®
098-1244006
300075-590EA 151.6 USD
300075-590
EZ Stop® III Shock Absorbing Lanyards, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Limits arresting forces to 900 lbs. or less
  • Soft cover shock absorber for comfort
  • 1-3/8" tubular web jacket and polyester core for durability and longevity