Uvex Classic™ Goggles, Honeywell Safety
Supplier: Honeywell Safety
763-S350CF
300047-432EA 44.42 USD
300047-432
Uvex Classic™ Goggles, Honeywell Safety
Lenses made of polycarbonate.
  • Fits over most Rx spectacles
  • Indirect hooded ventilation with foam
  • NOT for chemical splash applications
Weight: 0.15 lbs.