Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
Howard Leight QB3HYG® Banded Earplugs, Honeywell Safety
Supplier: ORS NASCO, INC.
These earplugs are made of polyurethane foam molded material.
  • Designed for people who are exposed to intermittent noise, QB3HYG offers good protection with its semi-aural fit
  • Smooth, ergonomic pods fit outside the opening of the ear canal for noise protection, while the patented band design prevents ear pods from touching dirty or contaminated surfaces when set down
  • Maximum attenuation of 42.400 dB at 8000.000 Hz
Certifications: Meets ANSI S.3.19-1974 standards.
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Color Noise Reduction Rating Packaging Weight Supplier No. VWR Catalog Number Unit Price Quantity
Orange/Yellow 23 dB Resealable Bag 0.05 lbs 154-QB3HYG 300007-189 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty