Next™ Skull Screws™ Earplugs, Peltor
Supplier: Peltor
Next™ Skull Screws™
247-P1300 247-P1301
300007-560PK 253.32 USD
300007-560 300007-559
Next™ Skull Screws™ Earplugs, Peltor
Earplugs Reusable Earplugs
These earplugs are made of polyurethane material.
  • Screws combine the tough look of threaded metal with the soft, comfortable feel of foam
  • Push-to-fit convenience
  • Combine comfort and noise attenuation for long-term wear