Stationary Electric-Driven Compressors, Ingersoll-Rand
Supplier: Ingersoll-Rand
383-20103172
300069-562EA 1476 USD
300069-562
Stationary Electric-Driven Compressors, Ingersoll-Rand
Pumps Compressors
Flow rate: 18.10 CFM @ 90 psi
  • 100% continuous duty cycle for the toughest applications and extended pump life for years of trouble-free service