Series 547 Digimatic Depth Gages, Mitutoyo
Supplier: Mitutoyo
504-547-218S
300082-218EA 690.12 USD
300082-218
Series 547 Digimatic Depth Gages, Mitutoyo
Measuring and Leveling Tools
Absolute encoder.
  • SPC conector
  • Measuring unit inch or metric
  • Digital display
Includes 4 extension rods.