Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Series 500 MyCal Solar Absolute Digimatic Calipers, Mitutoyo

Supplier: Mitutoyo
504-500-475 504-500-474
  • Eliminates the need for zero setting each time the caliper is turned on
  • Always turn on showing its true position
  • Memory of the origin point will not be lost when power is cut off
  • Provides unlimited response speed which means no more over-speed errors
  • For comparison measurements an incremental measurement function is also provided
  • Power is supplied by built-in solar cells which operate even in low light conditions down to 60 Lux



 

ORDER

Expand All / Collapse All
Inside Jaw Depth Outside Jaw Depth Measurement Range Supplier No. VWR Catalog Number Unit Price Quantity
0.65" 1.57" 0" [Min], 0 mm [Min], 6" [Max], 150 mm [Max] 504-500-474 300044-502 Each Retrieving
0.79" 2" 0" [Min], 0 mm [Min], 8" [Max], 200 mm [Max] 504-500-475 300043-838 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Accuracy ±0.001"
Includes Carring Case
Measuring System Inch/Metric
Body Material Stainless Steel
Display Type Digital LCD
Measurement Type Outside, Inside, Step, Depth
Resolution (Length) 0.000500", 0.010000 mm

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty