Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GSW Series Steel Rolling Warehouse Ladder w/ Handrails, Louisville Ladder®

Supplier: Louisville Ladder®
443-GSW2412 443-GSW2413 443-GSW2415 443-GSW2410 5011141172 443-GSW2406 443-GSW2408
  • Four spring loaded casters on 2-step to 5-step models
  • Casters retract when load is placed on the ladder, setting the ladder firmly in place on rubber tips
  • When the load is removed, springs push down the casters enabling the ladder to move about freely
  • Models 6-step and taller come equipped with a large 4" swivel caster in front and rigid casters in the rear
  • Stepping on bottom step activates the locking mechanism, locking rubber tipped feet in down position: To move the ladder, just step on the unlocking pedal
  • The top platform on models 12-step and taller is surrounded on three sides by a 4" high toeboard



 

ORDER

Expand All / Collapse All
Depth Height Width Platform Height No. of Steps Weight Capacity Supplier No. VWR Catalog Number Unit Price Quantity
50" [Max] 96" 34" [Max] 60" 6 109 lbs. 350 lbs. [Max] 443-GSW2406 300083-800 Each Retrieving
63" [Max] 116" 34" [Max] 80" 8 151 lbs. 350 lbs. [Max] 443-GSW2408 300083-802 Each Retrieving
77" [Max] 136" 34" [Max] 100" 10 198 lbs. 350 lbs. [Max] 443-GSW2410 300083-804 Each Retrieving
97" [Max] 166" 39" [Max] 130" 13 289 lbs. 350 lbs. [Max] 443-GSW2413 300083-808 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Material Steel
Step Depth 7"
Step/Rung Shape Flat
Platform Depth 10"
Step/Rung Width 24"

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty