Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TLS2200® Thermal Labels, Brady

Supplier: Brady Worldwide
TLS2200®
139838 262-PTL-21-427 139848 18391 11244 18680 11178 18458 18635 18416 18636
Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.

  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • Service temperature of –40 to 70 °C

Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text.

Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved.



 

ORDER

Expand All / Collapse All
Description Material For Use With Height Thickness Width Color Finish Print Area Packaging Supplier No. VWR Catalog Number Unit Price Quantity
TLS2200® Thermal Labels Polyester BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 0.004" 0.25" Clear Gloss Roll of 50 feet 11244 76243-666 Each Retrieving
TLS2200® Thermal Labels, Rectangle Polyimide BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 0.75" 0.0044" 0.25" White Gloss 0.25H×1.1"W 750 Labels/Roll 139838 76246-534 Each Retrieving
TLS2200® Thermal Labels Vinyl BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 21/2" 0.004" 1" White/Translucent Matte 0.75H×1"W 100 Labels/Roll 262-PTL-21-427 300065-918 Each Retrieving
TLS2200® Thermal Labels, Rectangle Polyvinylfluoride BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 2.5" 0.0022" 1" White Matte 0.75H×1"W 100 Labels/Roll 18636 76244-008 Each Retrieving
TLS2200® Thermal Labels, Rectangle Polyvinylfluoride BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 1.5" 0.0022" 1" White Matte 0.5H×1"W 250 Labels/Roll 18635 76244-682 Each Retrieving
TLS2200® Thermal Labels Vinyl BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System - 0.0042" 1" Red Gloss Roll of 50 Feet 18680 76251-604 Each Retrieving
TLS2200® Thermal Labels Vinyl BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 0.0042" 1" Green Gloss Roll of 50 Feet 11178 76251-606 Each Retrieving
TLS2200® Thermal Labels, Rectangle Polyester BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 0.5" 0.0044" 1.13" Light Gray Matte 0.5H×1.13"W 500 Labels/Roll 18391 76242-082 Each Retrieving
TLS2200® Thermal Labels Polyolefin BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 0.335" - 1.1677" White Matte 100 Labels/Roll 18458 76246-594 Each Retrieving
TLS2200® Thermal Labels, Rectangle Polyester BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 0.5" 0.0044" 1.5" Light Gray Matte 0.5H×1.5"W 500 Labels/Roll 18416 76241-614 Each Retrieving
TLS2200® Thermal Labels Polyimide BMP®61 Labeling System, BMP®71 Label Printer, BradyPrinter™ M611, TLS 2200® Thermal Labeling System, TLS-PC LINK™ Thermal Labeling System 0.25" 0.0044" 2" Amber Matte 100 Labels/Roll 139848 76242-342 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 300065-918
Material Self-Laminating Vinyl (B-427)

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty