Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Inch Pound/Centimeter Kilogram Dial Torque Wrenches, Proto®

Supplier: Proto®
577-6181F 577-6169F 577-6168F 577-6177F
  • Accurate to ± 4% of torque reading in both directions at 20% to 100% of full scale
  • Torsion drive design - accuracy not effected by hand position
  • Dual scale (English/metric) with clockwise and counterclockwise operation - fewer wrenches needed to complete job
  • Memory needles are standard on all wrenches except electric light models
  • Easy-to-read, high contrast scale



 

ORDER

Expand All / Collapse All
Material Overall Length Includes Measuring System Weight Head Type Locking Type Torque Increment Torque Setting Supplier No. VWR Catalog Number Unit Price Quantity
Alloy Steel 10.630" Blow Molded Case Inch/Metric 2.20 lb Square Ball 110.000 in lb, 11.700 N-m Dial 577-6181F 300027-322 Each Retrieving
Alloy Steel 10" Blow Molded Case Inch/Metric 1 lb Square Ball 0.500 in lb, 1.110 N-m Dial 577-6168F 300027-312 Each Retrieving
Alloy Steel 10.630" Blow Molded Case Inch/Metric 1.30 lb Square Ball 15.000 in lb, 1.560 N-m Dial 577-6177F 300027-318 Each Retrieving
Alloy Steel 10.630" Blow Molded Case Inch/Metric 1.40 lb Square Ball 11.000 in lb, 1.560 N-m Dial 577-6169F 300027-314 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type Torque
Description 300027-322 300027-312 300027-318 300027-314
Torque 600.00 in lb [Max], 384.60 N-m [Max] 30.00 in lb [Max], 4.27 N-m [Max] 250.00 in lb [Max], 160.30 N-m [Max] 75.00 in lb [Max], 10.70 N-m [Max]
Drive Size 3/8 in 1/4 in 3/8 in 1/4 in
Drive Type Male Square

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty