Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Inch Pound Ratchet Head Torque Wrenches, Proto®

Supplier: Proto®
577-6068C 577-6062C 577-6060A 577-6072C 577-6066C 577-6064C
  • Calibrated to +/- 4% in both the clockwise and counter clockwise
  • Calibration is accomplished by manual loading the torque wrenches
  • Ratcheting mechanisms are performance tested to over 80,000 cycles at 1/3 ANSI proof loads for durability
  • Manufactured to meet standard industrial torque applications, general industry, heavy equipment, and maintenance



 

ORDER

Expand All / Collapse All
Material Overall Length Finish Measuring System Weight Head Type Locking Type Torque Increment Torque Setting Supplier No. VWR Catalog Number Unit Price Quantity
Alloy Steel 11 7/8" Chrome - 2.55 lb - - 1.000 in lb - 577-6062C 300027-278 Each Retrieving
Alloy Steel 20 3/4" Chrome Inch 5.01 lb Ratchet Ball - - 577-6068C 300027-288 Each Retrieving
Alloy Steel 26 1/4" Chrome Inch 5 3/4 lb Ratchet Ball - - 577-6072C 300027-290 Each Retrieving
Alloy Steel 9 1/2" Chrome Inch 1.20 lb Pear Ball 1.000 in lb Micrometer 577-6060A 300027-276 Each Retrieving
Alloy Steel 11 7/8" Chrome - 2.63 lb - - 1.000 in lb - 577-6064C 300003-044 Each Retrieving
Alloy Steel 16 1/2" Chrome Inch 4.32 lb Ratchet Ball - - 577-6066C 300027-286 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type Torque
Description 300027-278 300027-288 300027-290 300027-276 300003-044 300027-286
Torque 40.00 in lb [Min], 200.00 in lb [Max] 300.00 in lb [Min], 1800.00 in lb [Max] 600.00 in lb [Min], 3000.00 in lb [Max] 10.00 in lb [Min], 50.00 in lb [Max] 40.00 in lb [Min], 200.00 in lb [Max] 100.00 in lb [Min], 1000.00 in lb [Max]
Drive Size 1/4 in 1/2 in 1/4 in 3/4 in 3/8 in
Drive Type Male Square

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty