Heavy-Duty Offset Striking Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
577-2642SW 577-2624SW 577-2637SW 577-2644SW 577-2633SW 577-2623SW 577-2630SW 577-2634SW 577-2641SW 577-2632SW 577-2620SW 577-2635SW 577-2628SW 577-2650SW 577-2648SW 577-2627SW 577-2649SW 577-2626SW 577-2636SW 577-2618SW 577-2621SW 577-2638SW 577-2629SW 577-2617SW 577-2647SW 577-2640SW
300024-294EA 221.57 USD
300024-294 300024-270 300024-290 300024-306 300024-326 300024-304 300024-268 300024-324 300024-288 300024-244 300024-300 300024-322 300024-264 300024-284 300024-240 300024-260 300024-280 300024-318 300002-836 300024-314 300024-310 300024-256 300024-330 300024-296 300024-252 300024-274
Heavy-Duty Offset Striking Wrenches, Proto®, ORS Nasco
Wrenches
Ideal for applications where extreme force is needed to loosen or set large fasteners.
  • Large striking surface
  • Heavy box walls provide greater torque transmission
  • Short handle permits work in close quarters
  • Offset provides clearance for knuckles and obstructions
  • Box opening remains on fastener when wrench is struck
  • Efficient transmittal of force to fastener