Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Foot Pound Ratchet Head Torque Wrenches, Proto®

Supplier: Proto®
577-6006C 577-6008C 577-6020AB 577-6014C 577-6016C 577-6018AB 577-6022B 577-6012C
  • Calibrated to +/- 3% clockwise direction and +/- 6% counter clockwise at 20% to 100% of full scale
  • Calibration is accomplished by manual loading the torque wrenches
  • Manufactured to meet standard industrial torque applications, general industry, heavy equipment, and maintenance



 

ORDER

Expand All / Collapse All
Material Overall Length Weight Head Type Locking Type Torque Increment Torque Setting Supplier No. VWR Catalog Number Unit Price Quantity
Alloy Steel 20 3/4" 3.97 lb Pear Ball 1.000 ft lb Micrometer 577-6016C 300003-040 Each Retrieving
Alloy Steel 16 7/16" 2 lb Pear Ball 0.500 ft lb Micrometer 577-6008C 300003-032 Each Retrieving
Alloy Steel 48" 14 1/2 lb Pear Ball 2.000 ft lb Micrometer 577-6022B 300027-224 Each Retrieving
Alloy Steel 26 1/4" 4.68 lb Pear Ball 1.000 ft lb Micrometer 577-6014C 300003-036 Each Retrieving
Alloy Steel 16 7/16" 2.40 lb Pear Ball 0.500 ft lb Micrometer 577-6012C 300003-034 Each Retrieving
Alloy Steel 41 1/2" 11.60 lb Pear Ball 2.000 ft lb Micrometer 577-6020AB 300003-042 Each Retrieving
Alloy Steel 16 7/16" 2 lb Pear Ball 0.500 ft lb Micrometer 577-6006C 300003-030 Each Retrieving
Alloy Steel 32 11/16" 9.70 lb Pear Ball 2.000 ft lb Micrometer 577-6018AB 300027-208 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type Torque
Description 300003-040 300003-032 300027-224 300003-036 300003-034 300003-042 300003-030 300027-208
Torque 30.00 ft lb [Min], 150.00 ft lb [Max] 10.00 ft lb [Min], 80.00 ft lb [Max] 140.00 ft lb [Min], 700.00 ft lb [Max] 50.00 ft lb [Min], 250.00 ft lb [Max] 20.00 ft lb [Min], 100.00 ft lb [Max] 90.00 ft lb [Min], 600.00 ft lb [Max] 10.00 ft lb [Min], 80.00 ft lb [Max] 60.00 ft lb [Min], 300.00 ft lb [Max]
Drive Size 1/2 in 1 in 1/2 in 3/8 in 3/4 in 3/8 in 3/4 in
Drive Type Male Square

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty