Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Click-Stop® Protoblack™ Adjustable Wrenches, Proto®

Supplier: Proto®
Click-Stop® Protoblack™
577-712SL 577-706SL 577-718SL 577-704SL 577-715SL 577-708SL 577-710SL
  • Extra wide jaw capacity
  • Large serrated knurl for jaw adjustment
  • Thin, slightly tapered jaw for access in tight work areas
  • Square jaw opening for greater strength
  • Clik-Stop® automatic locking jaw, locks jaw at any opening you select
  • Radius corner design increases strength and fastener surface contact area



 

ORDER

Expand All / Collapse All
Material Overall Length Finish Jaw Size Weight Handle Type Supplier No. VWR Catalog Number Unit Price Quantity
Forged Alloy Steel 4" Black Oxide 1/2" [Max] 0.14 lb I-Beam 577-704SL 300027-526 Each Retrieving
Forged Alloy Steel 10" Black Oxide 1 5/16" [Max] 0.84 lb I-Beam 577-710SL 300027-538 Each Retrieving
Forged Alloy Steel 15" Black Oxide 1 11/16" [Max] 3.22 lb I-Beam 577-715SL 300027-544 Each Retrieving
Forged Alloy Steel 12" Black Oxide 1 1/2" [Max] 142 lb I-Beam 577-712SL 300027-542 Each Retrieving
Forged Alloy Steel 6" Black Oxide 5/16" [Max] 0.28 lb I-Beam 577-706SL 300027-530 Each Retrieving
Forged Alloy Steel 18" Black Oxide 2 1/16" [Max] 5.60 lb I-Beam 577-718SL 300027-548 Each Retrieving
Forged Alloy Steel 8" Black Oxide 1 1/8" [Max] 0.52 lb I-Beam 577-708SL 300027-532 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type Adjustable
Head Angle 15°
Opening Type Jaw

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty