Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chrome Adjustable Wrenches, Crescent®

Supplier: Crescent®
181-AC18 181-AC110V 181-AC110 181-AC18V 181-AC112 181-AC14 181-AC16 181-AC124 181-AC14V 181-AC115 181-AC118
  • Wide capacity jaw (sizes 6-12) allows each tool to work with more fasteners sizes
  • Wider handles (sizes 6-12) provides more comfort and less stress to the hand
  • Tension spring stabilizes jaw and knurl adjusts easily
  • Alloy steel and heat treated
  • Proportioned dimensions for greatest strength and minimum weight
  • Chrome plated finish resists rust and corrosion
  • Polished head and regular, satin finished handle



 

ORDER

Expand All / Collapse All
Material Overall Length Finish Jaw Size Weight Handle Type Packing Type Supplier No. VWR Catalog Number Unit Price Quantity
Alloy Steel 8" Chrome 1 1/8" [Max] 9.01 oz I-Beam Boxed 181-AC18 300005-488 Each Retrieving
Alloy Steel 10" Chrome 1 5/16" [Max] 16.06 oz I-Beam Boxed 181-AC110 300001-876 Each Retrieving
Alloy Steel 12" Chrome 1 1/2" [Max] 25.97 oz I-Beam Boxed 181-AC112 300005-480 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type Adjustable
Head Angle 15°

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty