Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminum Pipe Wrenches, Proto®

Supplier: Proto®
577-810A 577-812A 577-818A 577-824A 577-836A 577-814A 577-848A
  • Graduation markings on hook jaw show approximate opening for pipe sizes
  • Large knurl and power threads provide fast, positive jaw adjustment
  • Light weight aluminum for less job fatigue
  • Replacement parts available
  • Standard handle lengths with wide gripping surface for increased comfort and one hand operation



 

ORDER

Expand All / Collapse All
Material Overall Length Jaw Size Measuring System Weight Supplier No. VWR Catalog Number Unit Price Quantity
Aluminum/Drop Forged Steel 36" 5" [Max] Inch 10.60 lb 577-836A 300000-559 Each Retrieving
Aluminum/Drop Forged Steel 48" 6" [Max] Inch 17 lb 577-848A 300000-561 Each Retrieving
Aluminum/Drop Forged Steel 24" 4" [Max] Inch 5.65 lb 577-824A 300000-557 Each Retrieving
Aluminum/Drop Forged Steel 12" 2" [Max] Inch 1.72 lb 577-812A 300000-551 Each Retrieving
Aluminum/Drop Forged Steel 18" 3 1/2" [Max] Inch 3.60 lb 577-818A 300000-555 Each Retrieving
Aluminum/Drop Forged Steel 10" 1 1/2" [Max] Inch 1.10 lb 577-810A 300000-549 Each Retrieving
Aluminum/Drop Forged Steel 14" 2" [Max] Inch 2.15 lb 577-814A 300000-553 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type Pipe
Head Angle 90°
Opening Type Jaw

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty