Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

12-Point Double End Flare Nut Wrenches, Proto®

Supplier: Proto®
577-3715MT 577-3713MT 577-3710MT 577-3709MT 577-3707MT 577-3719MT 577-3716MT
  • Modified square open end provides the strongest design with better fastener grip
  • Special alloy steel and heat treat permits thin box walls for strength and durability
  • Thin narrow jaws for better access in limited space



 

ORDER

Expand All / Collapse All
Material Head Width Overall Length Finish Head Thickness Jaw Size Measuring System Weight No. of Points Supplier No. VWR Catalog Number Unit Price Quantity
Forged Alloy Steel 28.800 mm, 31.200 mm 223.800 mm Chrome 11.700 mm, 13.200 mm 16 mm, 18 mm Metric 1.86 lb 12 577-3716MT 300024-786 Each Retrieving
Forged Alloy Steel 20.700 mm, 23.200 mm 160.300 mm Chrome 9.200 mm, 10 mm 9 mm, 11 mm Metric 1.01 lb 12 577-3709MT 300024-770 Each Retrieving
Forged Alloy Steel 25.700 mm, 27.600 mm 192 mm Chrome 10 mm, 11.700 mm 13 mm, 14 mm Metric 0.12 lb 12 577-3713MT 300024-780 Each Retrieving
Forged Alloy Steel 32.500 mm, 34.900 mm 239.700 mm Chrome 14 mm, 14.700 mm 19 mm, 21 mm Metric 3 lb 12 577-3719MT 300024-788 Each Retrieving
Forged Alloy Steel 17.200 mm, 18.900 mm 144.500 mm Chrome 7.500 mm, 8.300 mm 7 mm, 8 mm Metric 0.66 lb 12 577-3707MT 300024-766 Each Retrieving
Forged Alloy Steel 22 mm, 24.400 mm 176.300 mm Chrome 10 mm 7 mm, 8 mm Metric 1.26 lb 12 577-3710MT 300024-776 Each Retrieving
Forged Alloy Steel 28.200 mm, 30 mm 203 mm Chrome 11.700 mm, 12.500 mm 15 mm, 17 mm Metric 1.80 lb 12 577-3715MT 300024-784 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type Flare Nut

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty