Would you like to log in again?
Hang-On Cabinets, Kennedy
Supplier: Kennedy
444-306XB
  • Universal hang-on cabinets with front facing drawers make it easy to reach all your tools
  • All cabinets have internal drawer locking system and bolt to either side of your roller cabinet
  • Models 185 and 185X fit Kennedy models 273, 273X, 275, 275X, 277 and 277X
ORDER
Expand All / Collapse All
Material Depth Height Width Color No. of Drawers Drawer Dimensions Weight Supplier No. VWR Catalog Number Unit Price Quantity
Steel 20 in 33 1/8 in 13 5/8 in Brown Wrinkle 6 (1) 9-3/4"w x 18-1/2"d x 2"h, (4) 9-3/4"w x 18-1/2"d x 4"h, (1) 9-3/4"w x 18-1/2"d x 10"h 94 lb 444-306XB 300022-124 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Slide Type Ball-Bearing
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty