Plastic Pipe and Tubing Cutters, Ridgid®, ORS Nasco
Supplier: ORS Nasco
632-23493
300003-498EA 75.15 USD
300003-498
Plastic Pipe and Tubing Cutters, Ridgid®, ORS Nasco
Cutters
Cutters with X-CEL™ blade for fast single stroke cutting, no twisting or ratcheting required.
  • Durable construction for heavy duty use
Applicable on PVC, CPVC, PEX, PE, PB and Rubber Hose.