Pro Series Temperature and Humidity Data Logger, Dickson®
TP425
97043-472EA 89.94 USD
97043-472
Pro Series Temperature and Humidity Data Logger, Dickson®
Self-contained, economical data logger features an internal sensor for monitoring temperature and humidity in intervals ranging from one second to 24 hours
Its compact size allows it to be used in a wide range of environments. Current minimum and maximum data is displayed on the easy-to-read LCD screen. The internal memory stores more than 32,000 sample points, and the display flashes when the logger is full. Logger starts and stops with the push of a button. Features high speed USB connectivity and a user replaceable battery