jetPEI® Macrophage, Primary Macrophages, Glial and Dendritic Cells Transfection Reagent, Polyplus-transfection
Supplier: Polyplus-transfection
jetPEI®
103-05N
CA89129-974
jetPEI® Macrophage, Primary Macrophages, Glial and Dendritic Cells Transfection Reagent, Polyplus-transfection
Transfection Reagents
jetPEI®-Macrophage DNA transfection reagent is dedicated to the transfection of macrophage primary cells and cell lines which express mannose receptors, such as RAW 264.7.
  • Efficient transfection of primary macrophages, glial & dendritic cells
  • Easy-to-use protocol
  • Compatible with serum and antibiotics
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Every batch of jetPEI®-Macrophage is tested in a transfection assay. Typically, transfection of a firefly luciferase gene under the control of the CMV promote gives 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Delivery information: 150 mM NaCl solution is included. 0.5 ml of jetPEI®-Macrophage transfection reagent is sufficient to perform up to 250 transfections in 24-well plates.