    Sort by:
  • Search Within Results
4-Fluorobenzal chloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
Sort by:
Catalog Number: (TCF0315-025G)
Supplier:  TCI America
Description:   CAS Number: 456-19-9 MDL Number: MFCD00000832 Molecular Formula: C7H5Cl2F Molecular Weight: 179.02 Purity/Analysis Method: 95.0% (GC) Form: C...