Clamp Service Kit contains assorted sizes of zinc plated 2-Ear Clamps for industrial use and OEM applications
- Tamper-proof durable seal due to robust construction
- Ideal for air and fluid media lines
- Specially deburred edges reduce the risk of damage
- Tampering is visible
- Fast and simple installation
Suitable for EPDM rubber hose, plastic tubing, electrical cable, and high-vibration equipment hoses.
Ordering information: Kit includes "(15) of 5/16", (15) of 3/8", (15) of 1/2", (15) of 9/16", (15) of 5/8", (12) of 3/4", (12) of 7/8", (12) of 15/16", (12) of 1", (1) Standard Jaw Pincers"