Data Trol™ Control Serums, Thermo Scientific
Supplier: Thermo Scientific
Data Trol™
TR41001
46100-368EA 285.63 USD
46100-368
Data Trol™ Control Serums, Thermo Scientific
Control reagents
These serums are for monitoring the accuracy and precision of commonly used clinical chemistry assays
They have a 10-day stability after reconstitution, and require storage at 2 to 8°C