    Sort by:
  • Search Within Results
You Searched for: "1-benzyloxy-3-iodobenzene"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Benzyl 3-iodophenyl ether. Grade: 99. Melting Point C50-51*. Boiling Point C: NA. C13H11IO. 107623-21-2. LIGHT SENSITIVE