    Sort by:
  • Search Within Results
4-bromophenylboronic-acid-neopentyl-glycol-ester
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   2-(4-Bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane. Grade: 98+. Melting Point C109-111*. Boiling Point C: NA. C11H14BBrO2. 183677-71-6. MOISTURE SENSITIVE