TAT'L® Leak Detector, Shamrock
662-8OZ-TATL
300040-146CS 109.31 USD
300040-146
TAT'L® Leak Detector, Shamrock
  • Unique liquid leak detector with a special formula that stays where it is sprayed
  • Hundreds of bubbles are formed from the smallest leak
  • Leaks are detected instantly
  • Non-Corrosive
  • Non-Staining
  • Non-Toxic
  • pH value 6.4