Instant Super Degreaser, LPS
Supplier: LPS®
428-00720
300000-653CS 515.21 USD
300000-653
Instant Super Degreaser, LPS
Instant Super Degreaser removes grease, oil, wax, dirt, tar, and other contaminants.
  • Heavy-duty performance
  • Nonflammable
  • Fast evaporating
  • Nonconductive
  • No rinsing and no residue
  • Safe on metals
  • Inverted spray valve