Coleman® Ultimate® Coolers, Therapak®
56241G
89171-100EA 436.75 USD
89171-100
Coleman® Ultimate® Coolers, Therapak®
Rustproof, leak-resistant coolers are equipped with no-tilt drains and 5.08cm (2”) of insulation built into the lid
Coolers feature two-way handles for flexible and easy carrying. The 90.3L (82 qt.) model is supplied with wheels