Double Mailers, 95 kPa, Therapak®
Supplier: Therapak
Tour-Guard®
38549G 38551G
89171-040CS 360.19 USD
89171-040 89171-036
Double Mailers, 95 kPa, Therapak®
Mailers and Shippers
These versatile double mailers provide a safe environment for the transport of various laboratory specimens.
Each mailer comes with a 95kPa pressure tested inner receptacle, an outer fiberboard container and secure, locking Twist-n-Pull™ end caps.