Would you like to log in again?
Revco® ExF Series Ultra-Low Temperature Freezers, Thermo Scientific
Revco®
EXF24086ARAKCR EXF32086ARAK EXF60086VRAK EXF60086DRAKLN EXF40086DRAKLN EXF24086ARAK EXF24086A EXF60086VRAK EXF60086DRAKCO EXF32086VRAK EXF40086ARAKCO EXF40086ARAKCR EXF24086DRAKCR EXF40086A EXF32086DRAK EXF24086V EXF60086DRAK EXF24086VRAK EXF60086V EXF40086VRAK EXF40086ARAK EXF40086DRAKCR EXF32086A EXF32086DRAKCO EXF40086ARAKLN EXF32086ARAKCO EXF32086ARAKCR EXF32086DRAKCR EXF60086DRAKCR EXF60086V EXF32086V EXF32086DRAKLN EXF24086DRAK EXF24086DRAKLN EXF32086D EXF40086DRAKCO EXF32086DRAK EXF32086A EXF24086D EXF40086V EXF40086V EXF24086DRAKCO EXF60086DRAKCO EXF24086DRAKCR EXF24086ARAKCO EXF32086ARAKLN EXF24086ARAKLN EXF32086ARAKCO EXF60086D EXF40086ARAKCR EXF40086ARAKLN EXF40086DRAK EXF40086ARAKCO EXF32086ARAK EXF40086D EXF40086DRAKCO EXF40086DRAKCR EXF32086ARAKCR EXF32086ARAKLN EXF60086DRAK EXF32086VRAK EXF40086A EXF24086ARAKCR EXF40086VRAK EXF24086A EXF24086ARAKLN EXF32086DRAKCR EXF60086DRAKLN EXF24086DRAKCO EXF40086DRAKLN EXF24086ARAK EXF24086V EXF40086DRAK EXF24086DRAK EXF32086DRAKCO EXF24086VRAK EXF40086ARAK EXF24086ARAKCO EXF60086D EXF24086D EXF24086DRAKLN EXF32086DRAKLN EXF60086DRAKCR EXF40086D EXF32086V EXF32086D
89200-590EA 19456.99 USD InStock
89200-590 CA89200-574 89200-588 89200-630 CA89200-626 CA89200-568 89194-906 CA89200-588 89200-652 CA89200-578 89200-646 89200-602 89200-592 89194-918 89200-576 89194-910 89200-586 CA89200-572 89194-926 CA89200-584 89200-580 89200-604 CA89194-912 89200-642 89200-624 CA89200-640 89200-596 CA89200-598 CA89200-608 CA89194-926 CA89194-916 CA89200-620 89200-570 89200-614 89194-914 89200-648 CA89200-576 89194-912 89194-908 CA89194-922 89194-922 89200-636 CA89200-652 CA89200-592 89200-634 CA89200-618 89200-612 89200-640 89194-924 CA89200-602 CA89200-624 89200-582 CA89200-646 89200-574 89194-920 CA89200-648 CA89200-604 CA89200-596 89200-618 CA89200-586 89200-578 CA89194-918 CA89200-590 89200-584 CA89194-906 CA89200-612 89200-598 CA89200-630 CA89200-636 89200-626 89200-568 CA89194-910 CA89200-582 CA89200-570 CA89200-642 89200-572 CA89200-580 CA89200-634 CA89194-924 CA89194-908 CA89200-614 89200-620 89200-608 CA89194-920 89194-916 CA89194-914
Revco® ExF Series Ultra-Low Temperature Freezers, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Revco® ExF ultra-low temperature freezers are ideally suited for life science, university, government and academic accounts where accessible controls and at-a-glance monitoring are preferred. A centralized information center, mounted at eye level, includes a microprocessor control, monitoring system, and optional chart recorder designed for price sensitive sample protection without compromising performance. These -86°C freezers offer easy, four-button operation and a clear, intuitive user interface.
  • A simple temperature display and visual LED's, guide non-technical users through the set-point temperature and alarm parameter setting process
  • Alarm indicates high and low temperature, power failure, door ajar, hot condenser, probe failure and low battery for mind knowing your freezer is operating as specified
Construction features include: 5 interior compartment doors which minimizes cold air loss/warm air intrusion during door openings, reinforced stainless steel interior and shelves, triple point door gasket, heavy-gauge, cold-rolled steel cabinets with powder coat paint finish, 5 in
Backup systems inject liquid CO2 or LN2 when cabinet temperature warms to a preset level. Built-in backup systems are available on all ExF models. Choose from ether factory installed or field installed. Liquid flow shuts off automatically when the freezer door is opened.
Rack solutions are available for all ExF models. Choose from both side access or sliding drawer racks for 2" boxes, 3" boxes, standard microplates, and deep well plates.
Temperature recorders are 15.2cm (6") circular models with 7-day temperature recording capability. Sensaphone telephone dialing systems connect to remote alarm contacts. The 4-channel system monitors up to four inputs and dials up to four phone numbers until the warning message is received and acknowledged. The 8-channel system monitors up to eight inputs and dials up to eight phone numbers until the warning message is received and acknowledged. Battery back-up is standard. Six D-size alkaline batteries are required for backup.
Certifications: UL, cUL Listed - CE Marked.
Delivery information: Racks, back-up systems and other accessories sold separately. Revco® ExF freezers are supplied with manufacturer's parts and labor warranty (two-year warranty in US, five-year warranty in Canada). Upright freezers have five inner doors and include a four adjustable stainless steel shelves. All models include one 2.5cm (1") diameter access port with cap. See 89200-286 for accessory listings. Contact your sales representative for further information on all optional accessories available."
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: Thermo Scientific provides several service options, such as Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care packages. To learn more about these services, simply search "Thermo Cold Storage Technical Service" in the VWR.com search field. In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST™@vwr.com for further details.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty