    Sort by:
  • Search Within Results
2,4-Dichloro-5-isopropoxyaniline
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Grade: 98. Melting Point C40-42*. Boiling Point C: 110-112*/0. 2mm. C9H11Cl2NO. 41200-96-8. HARMFUL