n-Heptane ≥96%

Danger

AC61036-1000
200006-620EA 47.31 USD
200006-620
n-Heptane ≥96%
n-Heptane
Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –90.6 °C
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://us.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg

Order Now

Specification Test Results

Appearance Colorless Liquid
Assay ≥96 %
Water ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR